Assay Details
Gene Symbol: ERO1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005561264.2 | XP_005561321.1 | 1 - 2 | 330 | 65 | |
XM_015453791.1 | XP_015309277.1 | 1 - 2 | 330 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
