Assay Details
Gene Symbol: KANSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005570713.2 | XP_005570770.2 | 6 - 7 | 1004 | 58 | |
XM_005570714.2 | XP_005570771.1 | 7 - 8 | 1114 | 58 | ||
XM_005570716.2 | XP_005570773.1 | 6 - 7 | 1179 | 58 | ||
XM_015430859.1 | XP_015286345.1 | 5 - 6 | 1078 | 58 | ||
XM_015430860.1 | XP_015286346.1 | 7 - 8 | 1162 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
