Assay Details
Gene Symbol: HPS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005567727.2 | XP_005567784.1 | 7 - 8 | 1262 | 81 | |
XM_015457366.1 | XP_015312852.1 | 7 - 8 | 1171 | 81 | ||
XM_015457367.1 | XP_015312853.1 | 6 - 7 | 1137 | 81 | ||
XM_015457368.1 | XP_015312854.1 | 7 - 8 | 1254 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
