Assay Details
Gene Symbol: DPY30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005576171.2 | XP_005576228.1 | 3 - 4 | 229 | 118 | |
XM_005576172.2 | XP_005576229.1 | 3 - 4 | 229 | 118 | ||
XM_005576173.2 | XP_005576230.1 | 3 - 4 | 227 | 118 | ||
XM_005576174.2 | XP_005576231.1 | 3 - 4 | 230 | 118 | ||
XM_005576175.2 | XP_005576232.1 | 3 - 4 | 226 | 118 | ||
XM_005576176.2 | XP_005576233.1 | 3 - 4 | 192 | 118 | ||
XM_005576177.2 | XP_005576234.1 | 3 - 4 | 219 | 118 | ||
XM_015433826.1 | XP_015289312.1 | 3 - 4 | 298 | 118 | ||
XM_015433827.1 | XP_015289313.1 | 3 - 4 | 330 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
