Assay Details
Gene Symbol: PLA2G4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015441310.1 | XP_015296796.1 | 18 - 19 | 2601 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
