Assay Details
Gene Symbol: AHSA2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005575789.2 | XP_005575846.1 | 3 - 4 | 450 | 131 | |
XM_015433613.1 | XP_015289099.1 | 3 - 4 | 445 | 131 | ||
XM_015433614.1 | XP_015289100.1 | 3 - 4 | 392 | 131 | ||
XM_015433615.1 | XP_015289101.1 | 3 - 4 | 350 | 131 | ||
XM_015433616.1 | XP_015289102.1 | 3 - 4 | 286 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
