Assay Details
Gene Symbol: PSMB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005553359.2 | XP_005553416.1 | 2 - 3 | 759 | 105 | |
XM_005553360.2 | XP_005553417.1 | 2 - 3 | 1249 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
