Assay Details
Gene Symbol: EIF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287321.1 | NP_001274250.1 | 3 - 4 | 555 | 80 | |
GenBank mRNA | AB169765.1 | - | 3 - 4 | 555 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
