Assay Details
Gene Symbol: LOC102130734 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005570062.2 | XP_005570119.1 | 21 - 22 | 3062 | 96 | |
XM_005570063.2 | XP_005570120.1 | 21 - 22 | 2828 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
