Assay Details
Gene Symbol: UBE2I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005590842.2 | XP_005590899.1 | 2 - 3 | 324 | 76 | |
XM_015442653.1 | XP_015298139.1 | 2 - 3 | 189 | 76 | ||
XM_015442654.1 | XP_015298140.1 | 2 - 3 | 94 | 76 | ||
XM_015442655.1 | XP_015298141.1 | 2 - 3 | 344 | 76 | ||
XM_015442656.1 | XP_015298142.1 | 2 - 3 | 99 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
