Assay Details
Gene Symbol: ELAPOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283681.1 | NP_001270610.1 | 2 - 3 | 256 | 60 | |
XM_015452084.1 | XP_015307570.1 | 1 - 2 | 358 | 60 | ||
GenBank mRNA | AB169307.1 | - | 2 - 3 | 256 | 60 |

More Information
Set Membership: |
![]() ![]() |
