Assay Details
Gene Symbol: KLHL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001319454.1 | NP_001306383.1 | 1 - 2 | 529 | 109 | |
GenBank mRNA | AB168257.1 | - | 1 - 2 | 530 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
