Assay Details
Gene Symbol: LDAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005576467.2 | XP_005576524.1 | 3 - 4 | 388 | 146 | |
XM_005576468.2 | XP_005576525.1 | 4 - 5 | 473 | 146 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
