Assay Details
Gene Symbol: TNFAIP8L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005541929.2 | XP_005541986.1 | 1 - 2 | 590 | 123 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005541929.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
