Assay Details
Gene Symbol: CHRNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005562905.2 | XP_005562962.1 | 6 - 7 | 1097 | 65 | |
XM_015454609.1 | XP_015310095.1 | 5 - 6 | 4667 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
