Assay Details
Gene Symbol: TMIGD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005583336.2 | XP_005583393.1 | 2 - 3 | 181 | 118 | |
XM_005583337.2 | XP_005583394.1 | 2 - 3 | 123 | 118 | ||
XM_005583338.2 | XP_005583395.1 | 2 - 3 | 181 | 118 | ||
XM_015438128.1 | XP_015293614.1 | 1 - 2 | 172 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
