Assay Details
Gene Symbol: LYPD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005589472.2 | XP_005589529.1 | 2 - 3 | 253 | 103 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005589472.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
