Assay Details
Gene Symbol: TRIM16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005582958.2 | XP_005583015.1 | 3 - 4 | 1272 | 102 | |
XM_005582959.2 | XP_005583016.1 | 3 - 4 | 434 | 102 | ||
XM_015437663.1 | XP_015293149.1 | 4 - 5 | 1674 | 102 | ||
XM_015437664.1 | XP_015293150.1 | 5 - 6 | 1772 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
