Assay Details
Gene Symbol: ABR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015437391.1 | XP_015292877.1 | 6 - 7 | 858 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
