Assay Details
Gene Symbol: DSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005586971.2 | XP_005587028.1 | 11 - 12 | 2258 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
