Assay Details
Gene Symbol: STIL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005543450.2 | XP_005543507.1 | 3 - 4 | 296 | 94 | |
XM_005543451.2 | XP_005543508.1 | 3 - 4 | 296 | 94 | ||
XM_005543452.2 | XP_005543509.1 | 3 - 4 | 863 | 94 | ||
XM_005543453.2 | XP_005543510.1 | 3 - 4 | 863 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
