Assay Details
Gene Symbol: SLAMF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005541241.2 | XP_005541298.1 | 5 - 6 | 1232 | 139 | |
XM_005541242.2 | XP_005541299.1 | 4 - 5 | 925 | 139 | ||
GenBank mRNA | AB742520.1 | - | 5 - 6 | 862 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
