Assay Details
Gene Symbol: ALK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005576218.2 | XP_005576275.1 | 22 - 23 | 4462 | 154 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
