Assay Details
Gene Symbol: FAM234A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005590739.2 | XP_005590796.1 | 9 - 10 | 1319 | 92 | |
XM_005590740.2 | XP_005590797.1 | 9 - 10 | 1805 | 92 | ||
XM_005590741.2 | XP_005590798.1 | 9 - 10 | 1582 | 92 | ||
XM_005590742.2 | XP_005590799.1 | 9 - 10 | 1427 | 92 | ||
XM_005590745.2 | XP_005590802.1 | 9 - 10 | 1319 | 92 | ||
XM_005590746.2 | XP_005590803.1 | 9 - 10 | 1319 | 92 | ||
XM_015442247.1 | XP_015297733.1 | 10 - 11 | 1450 | 92 | ||
XM_015442248.1 | XP_015297734.1 | 11 - 12 | 1508 | 92 | ||
XM_015442249.1 | XP_015297735.1 | 7 - 8 | 1501 | 92 | ||
XM_015442250.1 | XP_015297736.1 | 9 - 10 | 1319 | 92 | ||
XM_015442251.1 | XP_015297737.1 | 9 - 10 | 1582 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
