Assay Details
Gene Symbol: IL11RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005581441.2 | XP_005581498.1 | 11 - 12 | 2934 | 142 | |
XM_015437051.1 | XP_015292537.1 | 11 - 12 | 2350 | 142 | ||
XM_015437052.1 | XP_015292538.1 | 11 - 12 | 2946 | 142 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
