Assay Details
Gene Symbol: PNPLA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005592919.2 | XP_005592976.1 | 6 - 7 | 801 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
