Assay Details
Gene Symbol: DLEC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005546656.2 | XP_005546713.1 | 1 - 2 | 1016 | 123 | |
XM_005546657.2 | XP_005546714.1 | 1 - 2 | 1016 | 123 | ||
XM_005546658.2 | XP_005546715.1 | 1 - 2 | 1017 | 123 | ||
XM_005546659.2 | XP_005546716.1 | 1 - 2 | 1018 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() |
