Assay Details
Gene Symbol: PITPNC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005584753.2 | XP_005584810.1 | 6 - 7 | 1369 | 105 | |
XM_005584754.2 | XP_005584811.1 | 6 - 7 | 2420 | 105 | ||
XM_005584755.2 | XP_005584812.1 | 6 - 7 | 629 | 105 | ||
XM_015438739.1 | XP_015294225.1 | 6 - 7 | 1369 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
