Assay Details
Gene Symbol: CEMIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005581918.1 | XP_005581975.1 | 4 - 5 | 1558 | 74 | |
XM_005581919.2 | XP_005581976.1 | 4 - 5 | 1563 | 74 | ||
XM_015437273.1 | XP_015292759.1 | 4 - 5 | 2620 | 74 | ||
XM_015437275.1 | XP_015292761.1 | 4 - 5 | 1067 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
