Assay Details
Gene Symbol: TMEFF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005573793.2 | XP_005573850.1 | 9 - 10 | 1723 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
