Assay Details
Gene Symbol: CHRNA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005554728.2 | XP_005554785.1 | 6 - 7 | 1675 | 118 | |
XM_005554729.2 | XP_005554786.1 | 4 - 5 | 3853 | 118 | ||
XM_015450269.1 | XP_015305755.1 | 7 - 8 | 2574 | 118 | ||
XM_015450270.1 | XP_015305756.1 | 8 - 9 | 1756 | 118 | ||
XM_015450271.1 | XP_015305757.1 | 6 - 7 | 2476 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
