Assay Details
Gene Symbol: NCBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005582533.2 | XP_005582590.1 | 11 - 12 | 1491 | 108 | |
XM_005582534.2 | XP_005582591.1 | 11 - 12 | 1480 | 108 | ||
XM_005582536.2 | XP_005582593.1 | 10 - 11 | 1365 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
