Assay Details
Gene Symbol: DGLUCY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005562016.2 | XP_005562073.2 | 12 - 13 | 1852 | 136 | |
XM_005562018.2 | XP_005562075.1 | 12 - 13 | 1974 | 136 | ||
XM_015454128.1 | XP_015309614.1 | 12 - 13 | 2224 | 136 | ||
XM_015454129.1 | XP_015309615.1 | 12 - 13 | 2184 | 136 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
