Assay Details
Gene Symbol: NEDD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005571925.2 | XP_005571982.1 | 2 - 3 | 331 | 79 | |
XM_005571927.2 | XP_005571984.1 | 2 - 3 | 226 | 79 | ||
XM_005571928.2 | XP_005571985.1 | 2 - 3 | 237 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
