Assay Details
Gene Symbol: ZNF598 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005590925.2 | XP_005590982.1 | 2 - 3 | 314 | 100 | |
XM_015442337.1 | XP_015297823.1 | 5 - 6 | 752 | 100 | ||
XM_015442338.1 | XP_015297824.1 | 5 - 6 | 753 | 100 | ||
XM_015442339.1 | XP_015297825.1 | 5 - 6 | 752 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
