Assay Details
Gene Symbol: MRPL52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005560807.2 | XP_005560864.2 | 4 - 5 | 271 | 88 | |
XM_005560808.2 | XP_005560865.2 | 4 - 5 | 271 | 88 | ||
XM_005560810.2 | XP_005560867.1 | 3 - 4 | 427 | 88 | ||
XM_005560811.2 | XP_005560868.1 | 2 - 3 | 149 | 88 | ||
XM_005560812.2 | XP_005560869.1 | 2 - 3 | 665 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
