Assay Details
Gene Symbol: APOBEC3H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015457180.1 | XP_015312666.1 | 1 - 2 | 332 | 124 | |
XM_015457181.1 | XP_015312667.1 | 1 - 2 | 334 | 124 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015457180.1, XM_015457181.1 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
