Assay Details
Gene Symbol: PRSS38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005541051.2 | XP_005541108.1 | 1 - 2 | 619 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
