Assay Details
Gene Symbol: NSUN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005543481.2 | XP_005543538.1 | 2 - 3 | 834 | 157 | |
XM_015436512.1 | XP_015291998.1 | 2 - 3 | 637 | 157 | ||
XM_015436513.1 | XP_015291999.1 | 3 - 4 | 639 | 157 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
