Assay Details
Gene Symbol: LOC102145105 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005543212.2 | XP_005543269.1 | 2 - 3 | 156 | 116 | |
XM_005543214.2 | XP_005543271.1 | 2 - 3 | 156 | 116 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005543212.2, XM_005543214.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
