Assay Details
Gene Symbol: POMC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005576403.2 | XP_005576460.1 | 2 - 3 | 548 | 140 | |
XM_005576404.2 | XP_005576461.1 | 2 - 3 | 288 | 140 | ||
XM_005576405.1 | XP_005576462.1 | 3 - 4 | 391 | 140 | ||
XM_005576406.1 | XP_005576463.1 | 3 - 4 | 340 | 140 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
