Assay Details
Gene Symbol: MYCL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005543857.2 | XP_005543914.1 | 3 - 4 | 780 | 144 | |
XM_005543858.2 | XP_005543915.1 | 1 - 2 | 516 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
