Assay Details
Gene Symbol: MYC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005564062.2 | XP_005564119.1 | 2 - 3 | 1374 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
