Assay Details
Gene Symbol: KCNJ9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005541276.2 | XP_005541333.1 | 3 - 4 | 2536 | 120 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005541276.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
