Assay Details
Gene Symbol: ELAPOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283681.1 | NP_001270610.1 | 6 - 7 | 805 | 77 | |
XM_015452080.1 | XP_015307566.1 | 6 - 7 | 951 | 77 | ||
XM_015452084.1 | XP_015307570.1 | 5 - 6 | 907 | 77 | ||
GenBank mRNA | AB169307.1 | - | 6 - 7 | 805 | 77 |
