Assay Details
Gene Symbol: TSC22D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005546083.2 | XP_005546140.1 | 1 | 541 | 134 | |
XM_005546084.2 | XP_005546141.1 | 1 | 541 | 134 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005546083.2, XM_005546084.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
