Assay Details
Gene Symbol: IL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005557731.2 | XP_005557788.2 | 1 - 2 | 837 | 80 | |
GenBank mRNA | DQ679797.1 | - | 1 - 2 | 128 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
