Assay Details
Gene Symbol: HYAL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005547191.2 | XP_005547248.1 | 3 - 4 | 1258 | 78 | |
XM_005547192.2 | XP_005547249.1 | 3 - 4 | 1098 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
