Assay Details
Gene Symbol: GDF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005565201.2 | XP_005565258.1 | 1 - 2 | 798 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
