Assay Details
Gene Symbol: PCED1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005570651.2 | XP_005570708.1 | 1 - 2 | 1336 | 100 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005570651.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
